EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Nitroarenes (selected). 1,6-Dinitropyrene.
PMID 15326686
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284327
1,6-Dinitropyrene, 98%
C16H8N2O4