EMAIL THIS PAGE TO A FRIEND
Gastrointestinal endoscopy
A secure and less-invasive new method for creation of an internal enteric fistula by using magnets as a therapeutic modality for pancreaticocystocutaneous fistula: a case report.
PMID 15332048