EMAIL THIS PAGE TO A FRIEND
Skin therapy letter
Ciclopirox (Loprox) gel for superficial fungal infections.
PMID 15334276
Abstract
Ciclopirox (Loprox) is a broad-spectrum antifungal medication that also has antibacterial and anti-inflammatory properties. Its main mode of action is thought to be its high affinity for trivalent cations, which inhibit essential co-factors in enzymes. Clinical trials have shown that ciclopirox gel is a successful treatment for seborrheic dermatitis of the scalp as well as for tinea pedis. Adverse effects are generally mild and include a skin-burning sensation, contact dermatitis, and pruritus. Ciclopirox is indicated in the US for the treatment of tinea pedis, tinea corporis, pityriasis versicolor, seborrheic dermatitis, and cutaneous candidiasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000040
Ciclopirox, European Pharmacopoeia (EP) Reference Standard
C12H17NO2