EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Selenium sulfide.
PMID 15334725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se