EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Electrophoretic analysis of di- and oligosaccharides derived from glycosaminoglycans on microchip format.
PMID 15351042
Abstract
Microchip electrophoresis is a powerful tool for fast analysis of nucleic acids and has expanded its applicability to the analysis of various biological materials including proteins and carbohydrates. Glycosaminoglycans have intrinsic negative charges, and are good targets for electrophoretic analysis. In the present paper, we developed a method to analyze oligosaccharides and unsaturated disaccharides derived from some glycosaminoglycans after digestion with specific enzymes followed by derivatization with 2-aminoacrydone (AMAC) by reductive amination. The method described here allowed rapid analysis of oligosaccharides derived from glycosaminoglycans within 150 s with high sensitivity. We show an application of the present technique to the glycosaminoglycan analysis in cultured HeLa cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06627
2-Aminoacridone, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
C13H10N2O