EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Neonatal administration of ACTH4-10 and its analog Semax to young laboratory mice modulates the number of catecholaminergic neurons in adult animal diencephalon.
PMID 15354819
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0401
Adrenocorticotropic Hormone Fragment 4-10 human, rat, ≥97% (HPLC)
C44H59N13O10S