EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
The remyelinating potential and in vitro differentiation of MOG-expressing oligodendrocyte precursors isolated from the adult rat CNS.
PMID 15355312