EMAIL THIS PAGE TO A FRIEND
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases
[Effect of 2,4-D isooctylester on sperms in tail of epididymis and HSP70 expression of testis tissue in rats].
PMID 15355714
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10615 2,4-D isooctyl ester, analytical standard