EMAIL THIS PAGE TO A FRIEND
Journal of toxicology. Clinical toxicology
Aphrodisiac drug-induced hemolysis.
PMID 15362601
Abstract
Volatile alkyl nitrites have been used during the past decades for "recreational purposes," and for intensifying sexual experience. Their use has been associated with methemoglobinemia and hemolysis. We report three patients who presented over the past year with acute hemolysis after inhalation of butyl nitrite, two of them had glucose-6-phosphate dehydrogenase (G6PD) deficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226637
Butyl nitrite, 95%
C4H9NO2