EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Pharmacological characterization and radioligand binding properties of a high-affinity, nonpeptide, bradykinin B1 receptor antagonist.
PMID 15363953
Abstract
Compound A (N-[2-[4-(4,5-dihydro-1H-imidazol-2-yl)phenyl]ethyl]-2-[(2R)-1-(2-napthylsulfonyl)-3-oxo-1,2,3,4-tetrahydroquinoxalin-2-yl]acetamide) is a member of a new class of aryl sulfonamide dihydroquinoxalinone bradykinin B1 receptor antagonists that should be useful pharmacological tools. Here we report on some of the pharmacological properties of compound A as well as the characterization of [35S]compound A as the first nonpeptide bradykinin B1 receptor radioligand. Compound A inhibited tritiated peptide ligand binding to the cloned human, rabbit, dog, and rat bradykinin B1 receptors expressed in CHO cells with Ki values of 0.016, 0.050, 0.56, and 29 nM, respectively. It was inactive at 10 microM in binding assays with the cloned human bradykinin B2 receptor. In functional antagonist assays with the cloned bradykinin B1 receptors, compound A inhibited agonist-induced signaling with activities consistent with the competition binding results, but had no antagonist activity at the bradykinin B2 receptor. Compound A was also found to be a potent antagonist in a rabbit aorta tissue bath preparation and to effectively block des-Arg9 bradykinin depressor responses in lipopolysaccharide-treated rabbit following intravenous administration. The binding of [35S]compound A was evaluated with the cloned bradykinin B1 receptors. In assays with human, rabbit, and dog receptors, [35S]compound A labeled a single site with Kd values of 0.012, 0.064, and 0.37 nM, respectively, and with binding site densities equivalent to those obtained using the conventional tritiated peptide ligands. Binding assays with the cloned rat bradykinin B1 receptor were not successful, presumably due to the low affinity of the ligand for this species receptor. There was no specific binding of the ligand detected in CHO cells expressing the human bradykinin B2 receptor. In assays with the cloned human bradykinin B1 receptor, the pharmacologies of the binding of [35S]compound A and [3H][Leu9]des-Arg10-kallidin were the same. The high signal-to-noise ratio obtained with [35S]compound A will allow this ligand to be a very useful tool for future investigations of the bradykinin B1 receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1542
[Lys-des-Arg9]-Bradykinin, ≥95% (HPLC)
C50H73N13O11