EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Regulation of serotonin 5-HT2C receptors by chronic ligand exposure.
PMID 15363976
Abstract
The effect of ligand pretreatment on human 5-hydroxytryptamine2C (5-HT2C) receptors was examined in CHO cells expressing high (CHO-1C7; 67+/-3 pmol/mg) or low (CHO-1C19; 72+/-10 fmol/mg) levels of the receptor. Seventy-two hours pretreatment of CHO-1C7 cells with various ligands did not affect receptor expression. Pretreatment with inverse agonists enhanced 5-HT-mediated inositol phosphate accumulation with no change in constitutive receptor activity. The enhanced agonist responsiveness was inversely correlated with the intrinsic activity of the pretreatment ligand. Seventy-two hours of pretreatment with the weak agonist, 5-methoxygramine, caused an elevation in constitutive activity but no alteration in 5-HT-mediated signaling. In CHO-1C19 cells, 24 but not 72 h of pretreatment with the inverse agonist mianserin enhanced 5-HT-mediated signaling, with no effect on basal signaling; pretreatment with 5-methoxygramine had no significant effect. These findings highlight differences in the pattern of chronic regulation of 5HT2C receptor signaling between high and low receptor expression levels in a common cellular background.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M14870
5-Methoxygramine, 99%
C12H16N2O
I5763
Indophenol
C12H9NO2