EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Treatment mostly not in line with current knowledge. New guidelines for sinusitis therapy].
PMID 15366497
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0037
Ceftibuten hydrate, ≥98% (HPLC)
C15H14N4O6S2 · xH2O