EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Migration of trimellitic acid from epoxy anhydride can coatings into foods.
PMID 15370844
Abstract
The migration of trimellitic acid and its esters from epoxy anhydride coatings was determined in simulants as well as in canned foods. The most appropriate simulant was a combination of EC simulants B and C: 2% acetic acid/10% ethanol in water. The average migration into food was 900 microg kg(-1). This far exceeds the 50 microg kg(-1) for which the safety of trimellitic acid and its anhydride is ensured and the Swiss legal limit (QM(T) of 5 mg kg(-1) coating). Furthermore, much trimellitic acid migrated as (unidentified) esters, i.e. toxicological testing of free trimellitic acid is inadequate for the material that in reality migrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4589
1,2,4-Benzenetricarboxylic acid, ≥99%
C9H6O6