EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Total synthesis of thiostrepton, part 2: construction of the quinaldic acid macrocycle and final stages of the synthesis.
PMID 15372630
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160660
Quinaldic acid, 98%
C10H7NO2