EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
The site-specific incorporation of p-iodo-L-phenylalanine into proteins for structure determination.
PMID 15378068
Abstract
A recently developed method makes it possible to genetically encode unnatural amino acids with diverse physical, chemical or biological properties in Escherichia coli and yeast. We now show that this technology can be used to efficiently and site-specifically incorporate p-iodo-L-phenylalanine (iodoPhe) into proteins in response to an amber TAG codon. The selective introduction of the anomalously scattering iodine atom into proteins should facilitate single-wavelength anomalous dispersion experiments on in-house X-ray sources. To illustrate this, we generated a Phe153 --> iodoPhe mutant of bacteriophage T4 lysozyme and determined its crystal structure using considerably less data than are needed for the equivalent experiment with cysteine and methionine. The iodoPhe residue, although present in the hydrophobic core of the protein, did not perturb the protein structure in any meaningful way. The ability to selectively introduce this and other heavy atom-containing amino acids into proteins should facilitate the structural study of proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8757
4-Iodo-L-phenylalanine
C9H10INO2