EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Fasciola hepatica infestation as a very rare cause of extrahepatic cholestasis.
PMID 15378799
Abstract
Fasciola hepatica, an endemic parasite in Turkey, is still a very rare cause of cholestasis worldwide. Through ingestion of contaminated water plants like watercress, humans can become the definitive host of this parasite. Cholestatic symptoms may be sudden but in some cases they may be preceded by a long period of fever, eosinophilia and vague gastrointestinal symptoms. We report a woman with cholangitis symptoms of sudden onset which was proved to be due to Fasciola hepatica infestation by an endoscopic retrograde cholangiography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31622
Bithionol, VETRANAL®, analytical standard
C12H6Cl4O2S