EMAIL THIS PAGE TO A FRIEND
Canadian journal of microbiology
Development of selective media for the isolation and enumeration of Alternaria species from soil and plant debris.
PMID 15381969
Abstract
A new semi-selective medium, acidified weak potato-dextrose agar (AWPDA) with Mertect (active ingredient: thiabendazole), was developed for the isolation and enumeration of Alternaria species from samples of soil and plant debris. The medium was selected based on growth inhibition tests against Alternaria and several other commonly encountered saprobic fungi utilizing three antifungal agents, Botran (active ingredient: dichloran), Bayleton (active ingredient: triadimefon), and Mertect, and two basal media, acidified potato-dextrose agar (APDA) and AWPDA. Botran inhibited growth of Rhizopus stolonifer moderately, but had little effect on Cladosporium cladosporoides, Fusarium oxysporum, Penicillium chrysogenum, or Trichoderma harzianum. Bayleton inhibited growth of R. stolonifer and C. cladosporoides severely, and inhibited growth of F. oxysporum, P. chrysogenum, and T. harzianum moderately. Mertect inhibited growth of C. cladosporoides, F. oxysporum, P. chrysogenum, and T. harzianum completely, but had little or moderate effect on R. stolonifer. All three antifungal agents inhibited growth of Alternaria species slightly or moderately. The combination of Bayleton and Mertect inhibited growth of all fungi severely. A comparison of recovery rates of Alternaria from soil and plant debris samples on AWPDA with Mertect and weak potato-dextrose agar (WPDA) revealed that Alternaria spp. accounted for 63.6%-81.0% of recovered fungal isolates on AWPDA with Mertect as compared to 0.6%-2.7% of recovered isolates on WPDA. The AWPDA medium with Mertect exhibited superior selective growth of Alternaria species from samples of soil and plant debris, and will be useful in studies where the recovery and enumeration of Alternaria species is necessary.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D67820
2,6-Dichloro-4-nitroaniline, 96%
C6H4Cl2N2O2
45435
Dichloran, PESTANAL®, analytical standard
C6H4Cl2N2O2