EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Cortactin associates with N-cadherin adhesions and mediates intercellular adhesion strengthening in fibroblasts.
PMID 15383621
Abstract
The regulation of N-cadherin-mediated intercellular adhesion strength in fibroblasts is poorly characterized; this is due, in part, to a lack of available quantitative models. We used a recombinant N-cadherin chimeric protein and a Rat 2 fibroblast, donor-acceptor cell model, to study the importance of cortical actin filaments and cortactin in the strengthening of N-cadherin adhesions. In wash-off assays, cytochalasin D (1 microM) reduced intercellular adhesion by threefold, confirming the importance of cortical actin filaments in strengthening of N-cadherin-mediated adhesions. Cortactin, an actin filament binding protein, spatially colocalized to, and directly associated with, nascent N-cadherin adhesion complexes. Transfection of Rat-2 cells with cortactin-specific, RNAi oligonucleotides reduced cortactin protein by 85% and intercellular adhesion by twofold compared with controls (P<0.005) using the donor-acceptor model. Cells with reduced cortactin exhibited threefold less N-cadherin-mediated intercellular adhesion strength compared with controls in wash-off assays using N-cadherin-coated beads. Immunoprecipitation and immunoblotting showed that N-cadherin-associated cortactin was phosphorylated on tyrosine residue 421 after intercellular adhesion. While tyrosine phosphorylation of cortactin was not required for recruitment to N-cadherin adhesions it was necessary for cadherin-mediated intercellular adhesion strength. Thus cortactin, and phosphorylation of its tyrosine residues, are important for N-cadherin-mediated intercellular adhesion strength.
Related Materials