EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Characterization of California rice field soils susceptible to delayed phytotoxicity syndrome.
PMID 15386164
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45687
Thiobencarb, PESTANAL®, analytical standard
C12H16ClNOS