EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Glycosylation of l,4:3,6-dianhydro-D-glucitol (isosorbide).
PMID 15388358
Abstract
Condensation of 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl-, 2,3,4-tri-O-acetyl-alpha-D-xylopyranosyl- and of 2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl bromides with l,4:3,6-dianhydro-D-glucitol under Koenigs-Knorr conditions, and using the Helferich modification of the reaction showed regioselectivity in glysosylation at C-5 of isosorbide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7562
Acetobromo-α-D-galactose, ≥93% (TLC)
C14H19BrO9