EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
The antiparasitic compound licochalcone a is a potent echinocytogenic agent that modifies the erythrocyte membrane in the concentration range where antiplasmodial activity is observed.
PMID 15388483
Abstract
The well-known antiparasitic compound licochalcone A is a potent membrane-active agent that transforms normal erythrocytes into echinocytes in parallel with the inhibition of growth of Plasmodium falciparum cultures, the in vitro antiplasmodial effect apparently being an indirect effect on the host cell. In vitro experiments with synchronous cultures demonstrate that inhibition of invasion is the principal mechanism of growth inhibition. The erythrocyte membrane-modifying effect was also transiently observed in vivo in mice after intravenous administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68783
Licochalcone A, ≥96.0% (HPLC)
C21H22O4