EMAIL THIS PAGE TO A FRIEND
Cell Death and Differentiation
Alkylphosphocholine-induced glioma cell death is BCL-X(L)-sensitive, caspase-independent and characterized by massive cytoplasmic vacuole formation.
PMID 15389288