EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Adrenergic blocking drugs; blocking of excitatory responses to epinephrine and adrenergic nerve stimulation with N-alkyl-N-(2-chloroethyl)-benzhydrylamines.
PMID 15400129
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A53605
Benzhydrylamine, 97%
C13H13N