EMAIL THIS PAGE TO A FRIEND
Journal of animal science
The effect of sulfaquinoxaline administration on metabolism of vitamin A and carotene and on hemoglobin young dairy calves.
PMID 15436344
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7382
Sulfaquinoxaline sodium salt, ≥92.5%
C14H11N4NaO2S