EMAIL THIS PAGE TO A FRIEND
Nature
Reactions of 9-anthraldehyde in sunlight.
PMID 15439159
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
278688
9-Anthracenecarboxaldehyde, 97%
C15H10O