EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Palladium-catalyzed copolymerization of ethene with acrolein dimethyl acetal: catalyst action and deactivation.
PMID 15453740
Abstract
Acrolein dimethyl acetal (ADMA) can be copolymerized with ethene using a cationic alpha-diimine palladium chelate catalyst to yield branched copolymers. Catalyst deactivation occurs via methanol elimination to give an inert eta3-1-methoxyallyl palladium species. This process can be retarded by the addition of an aliquot of methanol to the reaction mixture, increasing overall catalyst productivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190543
Acrolein dimethyl acetal, 98%
C5H10O2