EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Relationship between high sensitivity of suckling mice to hepatocarcinogenic and antiglucocorticoid effects of o-aminoazotoluene and diethylnitrosamine.
PMID 15455081
Abstract
Suckling mice were more sensitive to the hepatocarcinogenic effect of various carcinogens compared to adult animals. After treatment with o-aminoazotoluene and diethylnitrosamine HNF3-DNA-binding capacity and glucocorticoid-induced liver tyrosine aminotransferase activity in suckling mice decreased more significantly than in adult animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
121568
Fast Garnet GBC base, 97%
C14H15N3
31629
o-Aminoazotoluene, analytical standard
C14H15N3