EMAIL THIS PAGE TO A FRIEND
Antiviral chemistry & chemotherapy
Mechanisms of HIV receptor and co-receptor down-regulation by prostratin: role of conventional and novel PKC isoforms.
PMID 15457682
Abstract
Prostratin is an unusual non-tumour promoting phorbol ester with potential as an inductive adjuvant therapy for highly active antiretroviral therapy (HAART) due to its ability to up-regulate viral expression from latent provirus. In addition, prostratin is also able to inhibit de novo HIV infection most probably because it induces down-regulation of HIV receptors from the surface of target cells. In this study, we investigate the mechanisms by which prostratin down-regulates HIV receptor and co-receptor surface expression in lymphocytic and monocytic cell lines. Our results indicate that prostratin induces down-regulation of surface expression of CD4 and CXCR4, but not CCR5, in various cell lines. Down-regulation of CD4 and CXCR4 by prostratin is achieved by internalization through receptor-mediated endocytosis and/or macropinocytosis, which is then followed by degradation of these molecules. Because prostratin is a protein kinase C (PKC) activator, we next examined the potential contribution of distinct PKC isoforms to down-regulate CD4 and CXCR4 in response to prostratin stimulation. Although exposure of cells to prostratin or phorbol-myristate-acetate (PMA) induces the translocation of several PKC isoforms to the plasma membrane, the use of specific PKC inhibitors revealed that novel PKCs are the main mediators of the prostratin-induced CD4 down-regulation, whereas both conventional and novel PKCs contribute to CXCR4 down-regulation. Altogether these results showed that prostratin, through the activation of conventional and/or novel PKC isoforms, rapidly reduces cell surface expression of CD4 and CXCR4, but not CCR5, by inducing their internalization and degradation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0077
Prostratin, ≥98% (HPLC)
C22H30O6