EMAIL THIS PAGE TO A FRIEND
Biotechnology progress
Phases dispersion and oxygen transfer in a simulated fermentation broth containing castor oil and proteins.
PMID 15458353
Abstract
The sizes of air bubbles and castor oil drops were studied by image analysis as a function of the concentration of soluble protein (bovine serum albumin [BSA] and lipase, as model proteins) in a three-phase system using a simulated fermentation medium (aqueous salt solution, castor oil, and air). Small amounts of proteins (<0.02 g/L) caused an important decrease in oil drops and bubbles sizes, together with a pronounced decrease in surface tension. The extent and profiles of this decrease seem to be determined by the conformation of the protein at the interface. The kLa value increased considerably for increasing concentration (up to 0.02 g/L) of the two proteins but was very different (2-fold higher for the lipase) at the highest concentrations tested (0.5 g/L), a phenomenon that can be caused by the extent to which bubbles are trapped within oil drops.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W236004
γ-Decalactone, ≥98%, FCC, FG
C10H18O2
D804
γ-Decanolactone, 98%
C10H18O2