EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Comparative investigation of the effect of moclobemide and toloxatone on monoamine oxidase activity and psychometric performance in healthy subjects.
PMID 1546145
Abstract
The effects of moclobemide and toloxatone, two reversible monoamine oxidase-A inhibitors, on biochemical parameters that reflect monoamine metabolism and on psychomotor performance parameters were investigated in a study in 12 healthy volunteers. Treatments were given double-blind in a randomized, placebo-controlled cross-over design, with 1 week wash-out between the treatments. Drugs were given thrice daily in the following doses: moclobemide 150-150-150 mg and toloxatone 400-200-400 mg. All assessments were performed on day 8 under standardized conditions. There was no difference with regard to adverse events between moclobemide and toloxatone: both drugs induced a slight decrease in both supine and standing heart rate. Judged on the basis of the area under the curve, the two MAO-inhibitors reduced the plasma levels of DHPG and HVA, with more pronounced effects for moclobemide than for toloxatone. After moclobemide MAO-A inhibition was almost constant over 24 h, whereas the effect of toloxatone was short lasting after each dose. The same differences were reflected in plasma 5-HIAA concentrations and urinary excretion of 3-methoxytyramine and normetanephine. Neither of the compounds tested had any influence on the memory, vigilance, mood, or sleeping habits of the subjects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3452
Toloxatone, ≥98% (HPLC), solid
C11H13NO3