EMAIL THIS PAGE TO A FRIEND
Genome biology
A genome annotation-driven approach to cloning the human ORFeome.
PMID 15461802
Abstract
We have developed a systematic approach to generating cDNA clones containing full-length open reading frames (ORFs), exploiting knowledge of gene structure from genomic sequence. Each ORF was amplified by PCR from a pool of primary cDNAs, cloned and confirmed by sequencing. We obtained clones representing 70% of genes on human chromosome 22, whereas searching available cDNA clone collections found at best 48% from a single collection and 60% for all collections combined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5384 Aconitase from porcine heart