Brain research bulletin
Effects of NMDA glutamate receptor antagonist drugs on the volitional consumption of ethanol by a genetic drinking rat.
PMID 15464866
The ability of drugs that reduce NMDA receptor activity on the volitional consumption of ethanol in the genetic drinking rat, mHEP line, was investigated. After the consumption of ethanol solutions and water by each male or female mHEP rat had stabilized on its preferred concentration, different doses of LY 274614, a competitive NMDA antagonist, MK 801, a non-competitive NMDA antagonist, (+)-HA-966 or ACPC (1-aminocyclopropane-1-carboxylic acid), antagonists of the glycine site were administered daily for three days. The dose of 3.0 mg/kg i.p. LY 274614 reduced the consumption of ethanol by 64% compared to the pre-treatment baseline, while 0.3 mg/kg of MK 801 reduced consumption by 44%, 20 mg/kg (+)-HA-966 reduced consumption by 47% and 300 mg/kg of ACPC reduced consumption by 30%. These doses of LY 274614 and MK 801 reduced the ability of Sprague-Dawley rats to walk on a rotorod. Effects of these drugs on food intake were small except for the 20 mg/kg dose of (+)-HA-966. Therefore, the drugs did not have an anti-caloric effect and manipulations of the glutamatergic system through NMDA receptors may modify the consumption of ethanol. This interaction should be explored further for its therapeutic potential and to better understand the control by central neuronal systems of the consumption of ethanol.