EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Sulfone and phosphinic acid analogs of decaprenolphosphoarabinose as potential anti-tuberculosis agents.
PMID 15465326
Abstract
Mycobacteria biosynthesize a cell wall structure that is rich in polysaccharides containing arabinofuranose residues. The source of these arabinofuranose residues is decaprenolphosphoarabinose (1), the donor substrate for mycobacterial arabinosyltransferases. We have previously demonstrated that an analog of 1, C-phosphonate 7, prevented the growth of mycobacteria and this compound is currently undergoing testing for efficacy in tuberculosis-infected mice. We describe here the synthesis and testing of additional analogs of 1 that contain either a sulfone (8-14) or phosphinic acid (15-19) moiety in place of the phosphodiester functionality. Screening of these compounds in vitro against Mycobacterium tuberculosis strain H(37)Rv revealed that while some of these compounds possessed low to modest activity, none was as potent as 7.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W364703
3-Methyl-2-buten-1-ol, ≥98%, stabilized, FG
C5H10O
162353
3-Methyl-2-buten-1-ol, 99%
C5H10O