EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
GSK-3 phosphorylation of the Alzheimer epitope within collapsin response mediator proteins regulates axon elongation in primary neurons.
PMID 15466863