EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Effects of angiogenesis inhibitors on vascular network formation by human endothelial and melanoma cells.
PMID 15467037
Abstract
Endothelial cells involved in vasculogenesis and angiogenesis are key targets in cancer therapy. Recent evidence suggests that tumor cells can express some genes typically expressed by endothelial cells and form extracellular matrix-rich tubular networks, phenomena known as vasculogenic mimicry. We examined the effects of three angiogenesis inhibitors (i.e., anginex, TNP-470, and endostatin) on vasculogenic mimicry in human melanoma MUM-2B and C8161 cells and compared them with their effects in human endothelial HMEC-1 and HUVEC cells. Anginex, TNP-470, and endostatin markedly inhibited vascular cord and tube formation by HMEC-1 and HUVEC cells in vitro, whereas tubular network formation by MUM-2B and C8161 cells was relatively unaffected. Endothelial cells expressed higher mRNA and protein levels for two putative endostatin receptors, alpha5 integrin and heparin sulfate proteoglycan 2, than melanoma cells, suggesting a mechanistic basis for the differential response of the two cell types to angiogenesis inhibitors. These findings may contribute to the development of new antivascular therapeutic agents that target both angiogenesis and tumor cell vasculogenic mimicry.