EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Zirconium-mediated conversion of amides to nitriles: a surprising additive effect.
PMID 15468091
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293393
Hexanoamide, 98%
C6H13NO