EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Determination of sugar structures in solution from residual dipolar coupling constants: methodology and application to methyl beta-D-xylopyranoside.
PMID 15469309
Abstract
We have developed methodology for the determination of solution structures of small molecules from residual dipolar coupling constants measured in dilute liquid crystals. The power of the new technique is demonstrated by the determination of the structure of methyl beta-d-xylopyranoside (I) in solution. An oriented sample of I was prepared using a mixture of C(12)E(5) and hexanol in D(2)O. Thirty residual dipolar coupling constants, ranging from -6.44 to 4.99 Hz, were measured using intensity-based J-modulated NMR techniques. These include 15 D(HH), 4 (1)D(CH), and 11 (n)D(CH) coupling constants. The accuracy of the dipolar coupling constants is estimated to be < +/- 0.02 Hz. New constant-time HMBC NMR experiments were developed for the measurement of (n)D(CH) coupling constants, the use of which was crucial for the successful structure determination of I, as they allowed us to increase the number of fitted parameters. The structure of I was refined using a model in which the directly bonded interatom distances were fixed at their ab initio values, while 16 geometrical and 5 order parameters were optimized. These included 2 CCC and 6 CCH angles, and 2 CCCC and 6 CCCH dihedral angles. Vibrationally averaged dipolar coupling constants were used during the refinement. The refined solution structure of I is very similar to that obtained by ab initio calculations, with 11 bond and dihedral angles differing by 0.8 degrees or less and the remaining 5 parameters differing by up to 3.3 degrees . Comparison with the neutron diffraction structure showed larger differences attributable to crystal packing effects. Reducing the degree of order by using dilute liquid crystalline media in combination with precise measurement of small residual dipolar coupling constants, as shown here, is a way of overcoming the limitation of strongly orienting liquid crystals associated with the complexity of (1)H NMR spectra for molecules with more than 12 protons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5878
Methyl β-D-xylopyranoside, ≥99% (GC)
C6H12O5