EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric synthesis and translational competence of L-alpha-(1-cyclobutenyl)glycine.
PMID 15469317
Abstract
[reaction: see text] L-alpha-(1-Cyclobutenyl)glycine (1-Cbg) was targeted as a potentially translatable analogue of isoleucine and valine and as a useful building block for peptides. An enantioselective synthesis was executed in which the key step was diastereoselective addition of 1-cyclobutenylmagnesium bromide to the sulfinimine 2b derived from (S)-t-butanesulfinimide and tert-butyl glyoxylate. 1-Cbg was found to substitute efficiently for isoleucine and valine, but not leucine, in the translation of green fluorescent protein in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0377
DL-5-Hydroxylysine hydrochloride
C6H14N2O3 · HCl