EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Current applications of orcein in histochemistry. A brief review with some new observations concerning influence of dye batch variation and aging of dye solutions on staining.
PMID 15473577
Abstract
Current uses of orcein to demonstrate elastic fibers and, following permanganate oxidation (Shikata's modification), hepatitis B surface antigen, copper associated protein, and sulfated mucins, are reviewed. Variations in staining performance with batch of dye and age of dye solution is also discussed. Additional experimental findings support the view that the orcein stain for elastic tissue and Shikata's modification produces consistent, high quality results as long as appropriate controls and suitable dye batches, e.g., Biological Stain Commission certified dyes, are used.
Related Materials