EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Reversible and nonoxidative gamma-resorcylic acid decarboxylase: characterization and gene cloning of a novel enzyme catalyzing carboxylation of resorcinol, 1,3-dihydroxybenzene, from Rhizobium radiobacter.
PMID 15474471
Abstract
We found a gamma-resorcylic acid (gamma-RA, 2,6-dihydroxybenzoic acid) decarboxylase, as a novel enzyme applicable to carboxylation of resorcinol (RE, 1,3-dihydroxybenzene) to form gamma-RA, in a bacterial strain Rhizobium radiobacter WU-0108 isolated through the screening of gamma-RA degrading microorganisms. The activities for carboxylation of RE and decarboxylation of gamma-RA were detected in the cell-free extracts of R. radiobacter WU-0108 grown aerobically with gamma-RA. The enzyme, gamma-RA decarboxylase, was purified to homogeneity on SDS-PAGE through the steps of one ion-exchange chromatography and two kinds of hydrophobic chromatography. The molecular weight of the enzyme was estimated to be 130 kDa by gel-filtration, and that of the subunit was determined to be 34 kDa by SDS-PAGE, suggesting that the enzyme is a homotetrameric structure. The enzyme catalyzed the decarboxylation of gamma-RA, but not alpha-RA or beta-RA. Without addition of any cofactors, the enzyme catalyzed the regio-selective carboxylation of RE to form gamma-RA, without formation of alpha-RA and beta-RA, and of catechol to 2,3-dihydroxybenzoic acid. In the presence of oxygen, this gamma-RA decarboxylase showed no decrease in both of the activities as for decarboxylation of gamma-RA and carboxylation of RE, different from other decarboxylases reported so far. The gene, rdc, encoding the gamma-RA decarboxylase was cloned into Escherichia coli, sequenced, and subjected to over-expression. The deduced amino acid sequence of the rdc gene consists of 327 amino acid residues corresponding to 34 kDa protein, and shows 42% and 30% identity to those of a 2,3-dihydroxybenzoic acid decarboxylase from Aspergillus niger and a 5- carboxyvanillate decarboxylase from Sphingomonas paucimobilis SYK-6. A site-directed mutagenesis study revealed the two histidine residues at positions of 164 and 218 in Rdc to be essential for the catalytic activities of decarboxylation of gamma-RA and carboxylation of RE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D109606
2,6-Dihydroxybenzoic acid, 98%
C7H6O4