EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Well-defined calcium initiators for lactide polymerization.
PMID 15476371
Abstract
A series of monomeric amide or aryloxide complexes of the form LCaX, where L = CH[CMeNC(6)H(3)-2,6-](2), CH[CMeNC(6)H(4)-2-OMe](2), a bulky tris-pyrazolyl borate, Tp(iPr) or Tp(tBu) or 9-BBN-pz(2) and X = N(SiMe(3))(2) or OC(6)H(3)-2,6-, has been prepared and characterized and investigated in the ring-opening polymerizations of lactide. The compounds (Tp(tBu))CaX in THF are shown to be highly active and stereoselective. The propylene oxide complex (Tp(tBu))Ca(OC(6)H(3)-2,6-).(PO) has been isolated and structurally characterized (single-crystal X-ray) and shown to be inert to the polymerization of PO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
751979
Calcium iodide hydrate, 99.995% trace metals basis
CaI2 · xH2O