EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Antioxidant effect of hydroxytyrosol, a polyphenol from olive oil: scavenging of hydrogen peroxide but not superoxide anion produced by human neutrophils.
PMID 15476671
Abstract
Hydroxytyrosol (HT) (also known as dihydroxyphenylethanol (DPE)) is a polyphenol extracted from virgin olive oil. HT is known to exert an antioxidant effect but the mechanism of action and the identity of the reactive oxygen molecule(s) targeted are not known. In this study, we show that HT inhibits luminol-amplified chemiluminescence of human neutrophils stimulated with N-formyl-methionyl-leucyl-phenylalanine (fMLP), phorbol myristate acetate (PMA) and opsonized zymosan. This effect was dose-dependent and occurred immediately after the addition of HT. However, HT had no effect on lucigenin-amplified chemiluminescence, suggesting that it does not inhibit NADPH oxidase activation or scavenge superoxide anions. Furthermore, HT inhibited H(2)O(2)-dependent-dichlorofuoroscein (DCFH) fluorescence of activated neutrophils, as measured by flow cytometry. Finally, HT inhibited luminol-amplified chemiluminescence in a cell-free system consisting of H(2)O(2) and HRPO. These results suggest that HT, a polyphenol derived from olive oil, could exert its antioxidant effect by scavenging hydrogen peroxide but not superoxide anion released during the respiratory burst.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4291
3-Hydroxytyrosol, ≥98% (HPLC)
C8H10O3