EMAIL THIS PAGE TO A FRIEND
BMC pharmacology
In vitro pharmacokinetics of anti-psoriatic fumaric acid esters.
PMID 15479475
Abstract
Psoriasis is a chronic inflammatory skin disease that can be successfully treated with a mixture of fumaric acid esters (FAE) formulated as enteric-coated tablets for oral use. These tablets consist of dimethylfumarate (DMF) and salts of monoethylfumarate (MEF) and its main bioactive metabolite is monomethylfumarate (MMF). Little is known about the pharmacokinetics of these FAE. The aim of the present study was to investigate the hydrolysis of DMF to MMF and the stability of MMF, DMF and MEF at in vitro conditions representing different body compartments. DMF is hydrolyzed to MMF in an alkaline environment (pH 8), but not in an acidic environment (pH 1). In these conditions MMF and MEF remained intact during the period of analysis (6 h). Interestingly, DMF was hardly hydrolyzed to MMF in a buffer of pH 7.4, but was rapidly hydrolyzed in human serum having the same pH. Moreover, in whole blood the half-life of DMF was dramatically reduced as compared to serum. The concentrations of MMF and MEF in serum and whole blood decreased with increasing time. These data indicate that the majority of the FAE in the circulation are metabolized by one or more types of blood cells. Additional experiments with purified blood cell fractions resuspended in phosphate buffered saline (pH 7.4) revealed that at concentrations present in whole blood monocytes/lymphocytes, but not granulocytes and erythrocytes, effectively hydrolyzed DMF to MMF. Furthermore, in agreement with the data obtained with the pure components of the tablet, the enteric-coated tablet remained intact at pH 1, but rapidly dissolved at pH 8. Together, these in vitro data indicate that hydrolysis of DMF to MMF rapidly occurs at pH 8, resembling that within the small intestines, but not at pH 1 resembling the pH in the stomach. At both pHs MMF and MEF remained intact. These data explain the observation that after oral FAE intake MMF and MEF, but not DMF, can be readily detected in the circulation of human healthy volunteers and psoriasis patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C82604
Citraconic acid, 98%
C5H6O4
131040
Mesaconic acid, 99%
C5H6O4
128422
mono-Ethyl fumarate, 95%
C6H8O4