EMAIL THIS PAGE TO A FRIEND
Journal of virology
Effects on rotavirus cell binding and infection of monomeric and polymeric peptides containing alpha2beta1 and alphaxbeta2 integrin ligand sequences
PMID 15479820