EMAIL THIS PAGE TO A FRIEND
Pediatric radiology
Infantile Refsum disease: serial evaluation with MRI.
PMID 15480616
Abstract
Refsum disease is a rare metabolic disorder, which is characterized by the accumulation of phytanic acid in the blood and tissues, including the brain. A variant of this condition that occurs in young children is called infantile Refsum disease. The MRI findings of symmetrical signal change involving the corticospinal tracts, cerebellar dentate nuclei, and corpus callosum are characteristic. We report the serial MRI findings of a child with this rare metabolic disorder.