EMAIL THIS PAGE TO A FRIEND
Journal of Pineal Research
Comparison of target innervation by sympathetic axons in adult wild type and heterozygous mice for nerve growth factor or its receptor trkA.
PMID 15485548
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1542
Anti-Tyrosine Hydroxylase Antibody, clone, Chemicon®, from sheep