EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Protective role of Nd1 in doxorubicin-induced cardiotoxicity.
PMID 15485691
Abstract
The Ndl gene, which encodes a novel kelch family protein, is expressed ubiquitously in mouse tissues. In vitro studies suggest that Ndl protein, which binds to actin filaments, functions as a cytoskeletal stabilizer. In order to elucidate a physiological function of Ndl in vivo, we generated Nd1-deficient (Ndl-/-) mice. We developed Nd1-/- mice by standard gene targeting technique. Cardiac function was studied in wild type and Nd1-/- mice. Nd1-/- mice were viable and no gross anatomical abnormality was observed after birth. When mouse embryonic fibroblasts were cultured in the presence of cytochalasin D or doxorubicin, the number of apoptotic cells in the Nd1-/- cell culture was larger that that in the wild-type cell culture. Furthermore, Nd1-/- mice were sensitive to doxorubicin-induced cardiotoxicity with increased numbers of cardiomyocytes apoptosis. Although Nd1 is dispensable for normal mice development, Nd1 plays a protective role in doxorubicin-induced cardiotoxic responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44583
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
C27H29NO11 · HCl