EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Targeting heterogeneous nuclear ribonucleoparticule A1 and A2 proteins by RNA interference promotes cell death in transformed but not in normal mouse cell lines.
PMID 15486185