EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Comparison of three arginine-glycine-aspartate-containing peptides as inhibitors of equine platelet aggregation.
PMID 15500578