EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
High-throughput liquid chromatography-tandem mass spectrometry assay for plasma theophylline and its metabolites.
PMID 15502093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6885
1-Methyluric acid
C6H6N4O3
69720
1-Methylxanthine, ≥97.0% (HPLC)
C6H6N4O2
222526
3-Methylxanthine, 98%
C6H6N4O2